Ethyl-(S)-3-piperidinecarboxylate-D-tartrate ≥98.0% (by titrimetric analysis)
Proveedor: TCI
|
Atención
|
Sinónimos:
(S)-3-Piperidinecarboxylic acid ethyl ester D-Tartrate, Ethyl (S)-Nipecotate D-Tartrate, Ethyl (S)-3-Piperidinecarboxylate D-tartrate, (S)-Nipecotic acid ethyl ester D-Tartrate
N0681-25G
N0681-5G
TCIAN0681-25GEA
345
EUR
TCIAN0681-25G
TCIAN0681-5G
Ethyl-(S)-3-piperidinecarboxylate-D-tartrate ≥98.0% (by titrimetric analysis)
Ethyl-(S)-3-piperidinecarboxylate-D-tartrate
Formula:
C₁₂H₂₁NO₈ MW: 307,30 g/mol |
Número MDL:
MFCD03093639 Núm. CAS: 83602-38-4 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 155.0 to 158.0 deg-C |
Specific rotation [a]20/D | -9.5 to -11.0 deg (C=5, H2O) |
Solubility in Water | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...