3-(2-Naphthyl)-L-alanine hydrochloride ≥98.0% (by HPLC, titration analysis)

Proveedor: TCI

Sinónimos: H-2-Nal-OH·HCl, (S)-alpha-Amino-2-naphthalenepropionic Acid Hydrochloride

N0683-1G N0683-5G
TCIAN0683-1GEA 122 EUR
TCIAN0683-1G TCIAN0683-5G
3-(2-Naphthyl)-L-alanine hydrochloride ≥98.0% (by HPLC, titration analysis)
3-(2-Naphthyl)-L-alanine hydrochloride
Formula: C₁₃H₁₃NO₂·HCl
MW: 251,71 g/mol
Número MDL: MFCD03095595
Núm. CAS: 122745-12-4

Order Now

Resultados test especificaciones

Appearance White to Light yellow to Light orange powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Specific rotation [a]20/D -8.0 to -11.0 deg (C=1, 1mol/L HCl)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR