Dimethyl-2,7-naphthalenedicarboxylate ≥98.0% (por GC)
Proveedor: TCI
Sinónimos:
2,7-Naphthalenedicarboxylic Acid Dimethyl Ester
N0684-1G
N0684-5G
TCIAN0684-1GEA
190
EUR
TCIAN0684-1G
TCIAN0684-5G
Dimethyl-2,7-naphthalenedicarboxylate ≥98.0% (por GC)
Dimethyl-2,7-naphthalenedicarboxylate
Formula:
C₁₄H₁₂O₄ MW: 244,25 g/mol |
Número MDL:
MFCD03427560 Núm. CAS: 2549-47-5 |
Resultados test especificaciones
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 136.0 to 140.0 deg-C |
Solubility in hot Methanol | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...