6-Nitroanthranilic acid ≥97.0% (by HPLC, titration analysis)

Proveedor: TCI
Atención

N0818-1G N0818-5G
TCIAN0818-1GEA 16 EUR
TCIAN0818-1G TCIAN0818-5G
6-Nitroanthranilic acid ≥97.0% (by HPLC, titration analysis)
6-Nitroanthranilic acid
Formula: C₇H₆N₂O₄
Número MDL: MFCD00034785
Núm. CAS: 50573-74-5

Order Now

Resultados test especificaciones

Appearance Light yellow to Brown powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (Neutralization titration) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR