4-Nitrobenzyl-(2S,4S)-2-(dimethylcarbamoyl)-4-mercapto-1-pyrrolidinecarboxylate ≥95.0% (by HPLC, titration analysis)

Proveedor: TCI

Sinónimos: (2S,4S)-4-Nitrobenzyl 2-(dimethylcarbamoyl)-4-mercaptopyrrolidine-1-carboxylate, (2S,4S)-2-(Dimethylcarbamoyl)-4-mercapto-1-pyrrolidinecarboxylic Acid 4-Nitrobenzyl Ester, 4-Nitrobenzyl (2S,4S)-2-(Dimethylcarbamoyl)-4-mercapto-1-pyrrolidinecarboxylate

N0828-1G N0828-5G
TCIAN0828-1GEA 79 EUR
TCIAN0828-1G TCIAN0828-5G
4-Nitrobenzyl-(2S,4S)-2-(dimethylcarbamoyl)-4-mercapto-1-pyrrolidinecarboxylate ≥95.0% (by HPLC, titration analysis)
4-Nitrobenzyl-(2S,4S)-2-(dimethylcarbamoyl)-4-mercapto-1-pyrrolidinecarboxylate
Formula: C₁₅H₁₉N₃O₅S
Número MDL: MFCD08063348
Núm. CAS: 96034-64-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f3abc1b3177ab43efb22fe97481e4f3583139f3e&vid=f3abc1b3177ab43efb22fe97481e4f3583139f3e&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 95.0 area%
Purity (Iodometric Titration) min. 95.0 %
Melting point 117.0 to 121.0 deg-C
Specific rotation [a]20/D +8.0 to +10.0 deg (C=1, CHCl3)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR