Norlaudanosine hydrochloride ≥98.0% (by HPLC, total nitrogen)
Proveedor: TCI
|
Atención
|
Sinónimos:
1-(3,4-Dimethoxybenzyl)-6,7-dimethoxy-1,2,3,4-tetrahydroisoquinoline Hydrochloride, Tetrahydropapaverine Hydrochloride
N0918-5G
N0918-25G
TCIAN0918-5GEA
37
EUR
TCIAN0918-5G
TCIAN0918-25G
Norlaudanosine hydrochloride ≥98.0% (by HPLC, total nitrogen)
Norlaudanosine hydrochloride
Formula:
C₂₀H₂₅NO₄ MW: 379,88 g/mol |
Número MDL:
MFCD00035267 Núm. CAS: 6429-04-5 EINECS: 229-213-9 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Melting point | 216.0 to 220.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...