7-Nitro-1,2,3,4-tetrahydroquinoline ≥98.0% (by GC, titration analysis)
Proveedor: TCI
|
Atención
|
Sinónimos:
1,2,3,4-Tetrahydro-7-nitroquinoline
N0955-5G
N0955-25G
TCIAN0955-5GEA
25
EUR
TCIAN0955-5G
TCIAN0955-25G
7-Nitro-1,2,3,4-tetrahydroquinoline ≥98.0% (by GC, titration analysis)
7-Nitro-1,2,3,4-tetrahydroquinoline
Formula:
C₉H₁₀N₂O₂ MW: 178,19 g/mol |
Número MDL:
MFCD00496654 Núm. CAS: 30450-62-5 |
Resultados test especificaciones
Appearance | Light yellow to Brown powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 61.0 to 65.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...