S-[4-[(4-Nitrobenzyl)oxy]phenethyl]isothiourea methanesulphonate ≥98.0% (by HPLC)

Proveedor: TCI

Sinónimos: KB-R 7943, 4-((4-Nitrobenzyl)oxy)phenethyl carbamimidothioate methanesulphonate, Carbamimidothioic acid 4-[(4-Nitrobenzyl)oxy]phenethyl Ester Methanesulphonate, 4-[(4-Nitrobenzyl)oxy]phenethyl Carbamimidothioate Methanesulphonate

N0966-10MG N0966-50MG
TCIAN0966-10MGEA 68 EUR
TCIAN0966-10MG TCIAN0966-50MG
S-[4-[(4-Nitrobenzyl)oxy]phenethyl]isothiourea methanesulphonate ≥98.0% (by HPLC)
S-[4-[(4-Nitrobenzyl)oxy]phenethyl]isothiourea methanesulphonate
Formula: C₁₆H₁₇N₃O₃S
MW: 427,49 g/mol
Temperatura de almacenaje: Nevera
Número MDL: MFCD00952138
Núm. CAS: 182004-65-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0d4fd21e9a6e86e3e90de691e0fe7a9078c550d8&vid=0d4fd21e9a6e86e3e90de691e0fe7a9078c550d8&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Yellow to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Elemental analysis (Nitrogen) 9.50 to 10.30 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR