Dimethylammonium dichlorotri(mu-chloro)bis[(S)-(-)-2,2'-bis(diphenylphosphino)-1,1'-binaphthyl]diruthenate(II)

Proveedor: TCI

Sinónimos: [NH2Me2][{RuCl((S)-binap)}2(mu-Cl)3]

N1016-200MG N1016-1G
TCIAN1016-200MGEA 32 EUR
TCIAN1016-200MG TCIAN1016-1G
Dimethylammonium dichlorotri(mu-chloro)bis[(S)-(-)-2,2'-bis(diphenylphosphino)-1,1'-binaphthyl]diruthenate(II)
Dimethylammonium dichlorotri(mu-chloro)bis[(S)-(-)-2,2'-bis(diphenylphosphino)-1,1'-binaphthyl]diruthenate(II)
Formula: C₉₀H₇₂Cl₅NP₄Ru₂
Número MDL: MFCD09753033
Núm. CAS: 199541-17-8

Order Now

Resultados test especificaciones

Appearance Light yellow to Amber to Dark green powder to crystal
IR to pass test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR