Dimethylammonium dichlorotri(mu-chloro)bis[(R)-(+)-2,2'-bis(di-p-tolylphosphino)-1,1'-binaphthyl]diruthenate(II)

Proveedor: TCI
Peligro

Sinónimos: [NH2Me2][{RuCl((R)-tolbinap)}2(mu-Cl)3]

N1017-200MG N1017-1G
TCIAN1017-200MGEA 31 EUR
TCIAN1017-200MG TCIAN1017-1G
Dimethylammonium dichlorotri(mu-chloro)bis[(R)-(+)-2,2'-bis(di-p-tolylphosphino)-1,1'-binaphthyl]diruthenate(II)
Dimethylammonium dichlorotri(mu-chloro)bis[(R)-(+)-2,2'-bis(di-p-tolylphosphino)-1,1'-binaphthyl]diruthenate(II)
Formula: C₉₈H₈₈Cl₅NP₄Ru₂
MW: 1783,07 g/mol
Número MDL: MFCD09753034
Núm. CAS: 749935-02-2

Order Now

Resultados test especificaciones

Appearance Light yellow to Amber to Dark green powder to crystal
IR to pass test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR