Dimethylammonium dichlorotri(mu-chloro)bis{(S)-(-)-2,2'-bis[di(3,5-xylyl)phosphino]-1,1'-binaphthyl}diruthenate(II)

Proveedor: TCI

Sinónimos: [NH2Me2][(RuCl((S)-xylbinap))2(μ-Cl)3]

N1020-200MG N1020-1G
TCIAN1020-200MGEA 36 EUR
TCIAN1020-200MG TCIAN1020-1G
Dimethylammonium dichlorotri(mu-chloro)bis{(S)-(-)-2,2'-bis[di(3,5-xylyl)phosphino]-1,1'-binaphthyl}diruthenate(II)
Dimethylammonium dichlorotri(mu-chloro)bis{(S)-(-)-2,2'-bis[di(3,5-xylyl)phosphino]-1,1'-binaphthyl}diruthenate(II)
Formula: C₁₀₆H₁₀₄Cl₅NP₄Ru₂
MW: 1895,29 g/mol
Número MDL: MFCD09753025
Núm. CAS: 944451-10-9

Order Now

Resultados test especificaciones

Appearance Orange to Brown powder to crystal
IR to pass test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR