Dimethylammonium dichlorotri(mu-chloro)bis[(R)-(+)-5,5'-bis(diphenylphosphino)-4,4'-bi-1,3-benzodioxole]diruthenate(II)

Proveedor: TCI
Peligro

Sinónimos: [NH2Me2][{RuCl((R)-segphos®)}2(mu-Cl)3]

N1021-200MG N1021-1G
TCIAN1021-200MGEA 36 EUR
TCIAN1021-200MG TCIAN1021-1G
Dimethylammonium dichlorotri(mu-chloro)bis[(R)-(+)-5,5'-bis(diphenylphosphino)-4,4'-bi-1,3-benzodioxole]diruthenate(II)
Dimethylammonium dichlorotri(mu-chloro)bis[(R)-(+)-5,5'-bis(diphenylphosphino)-4,4'-bi-1,3-benzodioxole]diruthenate(II)
Formula: C₇₈H₆₄Cl₅NO₈P₄Ru₂
MW: 1646,65 g/mol
Número MDL: MFCD09753037
Núm. CAS: 346457-41-8

Order Now

Resultados test especificaciones

Appearance Orange to Brown powder to crystal
IR to pass test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR