Dimethylammonium dichlorotri(mu-chloro)bis[(S)-(-)-5,5'-bis(diphenylphosphino)-4,4'-bi-1,3-benzodioxole]diruthenate(II)
Proveedor: TCI
|
Peligro
|
Sinónimos:
[NH2Me2][{RuCl((S)-segphos®)}2(mu-Cl)3]
N1022-200MG
N1022-1G
TCIAN1022-200MGEA
33
EUR
TCIAN1022-200MG
TCIAN1022-1G
Dimethylammonium dichlorotri(mu-chloro)bis[(S)-(-)-5,5'-bis(diphenylphosphino)-4,4'-bi-1,3-benzodioxole]diruthenate(II)
Dimethylammonium dichlorotri(mu-chloro)bis[(S)-(-)-5,5'-bis(diphenylphosphino)-4,4'-bi-1,3-benzodioxole]diruthenate(II)
Formula:
C₇₈H₆₄Cl₅NO₈P₄Ru₂ MW: 1646,65 g/mol |
Número MDL:
MFCD09753037 Núm. CAS: 488809-34-3 |
Resultados test especificaciones
Appearance | Orange to Brown powder to crystal |
IR | to pass test |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...