Diamond nanoparticles 5% dispersion in water (<10 nm)

Proveedor: TCI

N1082-5G N1082-25G
TCIAN1082-5GEA 67 EUR
TCIAN1082-5G TCIAN1082-25G
Diamond nanoparticles 5% dispersion in water (<10 nm)
Diamond

Nanodiamonds (Nanocarbon Materials)

Número MDL: MFCD00211867
Núm. CAS: 7782-40-3
EINECS: 231-953-2
Índice Merck: 14,02987

Order Now

Resultados test especificaciones

Appearance Light gray to Brown to Black clear liquid to cloudy liquid
Concentration 4.5 to 5.4 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR