β-Nicotinamide mononucleotide ≥98.0% (by HPLC)
Proveedor: TCI
Sinónimos:
((2R,3S,4R,5R)-5-(3-Carbamoylpyridin-1-ium-1-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methyl hydrogen phosphate, beta-NMN, beta-Nicotinamide mononucleotide, beta-Nicotinamide Ribose monophosphate, β-Nicotinamide ribose monophosphate
N1123-50MG
N1123-250MG
TCIAN1123-50MGEA
62
EUR
TCIAN1123-50MG
TCIAN1123-250MG
β-Nicotinamide mononucleotide ≥98.0% (by HPLC)
β-Nicotinamide mononucleotide
Formula:
C₁₁H₁₅N₂O₈P MW: 334,22 g/mol Temperatura de almacenaje: Congelador |
Número MDL:
MFCD00038748 Núm. CAS: 1094-61-7 EINECS: 214-136-5 Índice Merck: 14,06663 |
Resultados test especificaciones
Appearance | White to Light yellow powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...