Notoginsenoside R1 ≥98.0% (by HPLC)

Proveedor: TCI

Sinónimos: Notoginsenoside R1, (3beta,6alpha,12beta)-20-(beta-D-Glucopyranosyloxy)-3,12-dihydroxydammar-24-en-6-yl 2-O-beta-D-Xylopyranosyl-beta-D-glucopyranoside, (2S,3R,4S,5S,6R)-2-(((S)-2-((3S,5R,6S,8R,9R,10R,12R,13R,14R,17S)-6-(((2R,3R,4S,5S,6R)-4,5-Dihydroxy-6-(hydroxymethyl)-3-(((2S,3R,4S,5R)-3,4,5-trihydroxytetrahydro-2H-pyran-2-yl)oxy)tetrahydro-2H-pyran-2-yl)oxy)-3,12-dihydroxy-4,4,8,10,14-pentamethylhexadecahydro-1H-cyclopenta[a]phenanthren-17-yl)-6-methylhept-5-en-2-yl)oxy)-6-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol, Sanchinoside R1

N1129-10MG N1129-50MG
TCIAN1129-10MGEA 69 EUR
TCIAN1129-10MG TCIAN1129-50MG
Notoginsenoside R1 ≥98.0% (by HPLC)
Notoginsenoside R1
Formula: C₄₇H₈₀O₁₈
MW: 933,14 g/mol
Temperatura de almacenaje: Nevera
Número MDL: MFCD00210535
Núm. CAS: 80418-24-2

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (HPLC) (CAD) min. 98.0 area%
Melting point 213.0 to 217.0 deg-C
Specific rotation [a]20/D +18.0 to +22.0 deg (C=0.5, MeOH)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR