3-Nitro-2-pyridinethiol ≥98.0% (by HPLC, titration analysis)

Proveedor: TCI

Sinónimos: 3-Nitropyridine-2(1H)-thione, 3-Nitropyridine-2-thiol

N1149-1G N1149-5G
TCIAN1149-1GEA 63 EUR
TCIAN1149-1G TCIAN1149-5G
3-Nitro-2-pyridinethiol ≥98.0% (by HPLC, titration analysis)
3-Nitro-2-pyridinethiol
Formula: C₅H₄N₂O₂S
MW: 156,16 g/mol
Número MDL: MFCD00661369
Núm. CAS: 38240-29-8

Order Now

Resultados test especificaciones

Appearance Light yellow to Brown powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 172.0 to 176.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR