Diethyl oxalacetate sodium salt ≥95.0% (by titrimetric analysis)

Proveedor: TCI
Atención

Sinónimos: Sodium diethyl oxalacetate, Sodium Diethyl Oxalacetate, Oxalacetic Acid Diethyl Ester Sodium Salt

O0074-25G O0074-500G
TCIAO0074-25GEA 26 EUR
TCIAO0074-25G TCIAO0074-500G
Diethyl oxalacetate sodium salt ≥95.0% (by titrimetric analysis)
Diethyl oxalacetate sodium salt
Formula: C₈H₁₂O₅
Número MDL: MFCD00035571
Núm. CAS: 40876-98-0
EINECS: 255-122-9

Order Now

Resultados test especificaciones

Appearance White to Light yellow to Light orange powder to crystal
Purity (Neutralization titration) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR