Diphenyl oxalate ≥98.0% (por GC)

Proveedor: TCI
Atención

Sinónimos: Oxalic Acid Diphenyl Ester

O0111-25G O0111-500G O0111-100G
TCIAO0111-25GEA 79 EUR
TCIAO0111-25G TCIAO0111-500G TCIAO0111-100G
Diphenyl oxalate ≥98.0% (por GC)
Diphenyl oxalate
Formula: C₁₄H₁₀O₄
MW: 242,23 g/mol
Número MDL: MFCD00059682
Núm. CAS: 3155-16-6

Order Now

Resultados test especificaciones

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 136.0 to 140.0 deg-C
Solubility in Acetone almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR