5-Oxoazelaic acid ≥96.0% (by GC, titration analysis)

Proveedor: TCI
Atención

O0258-1G O0258-5G
TCIAO0258-1GEA 170 EUR
TCIAO0258-1G TCIAO0258-5G
5-Oxoazelaic acid ≥96.0% (by GC, titration analysis)
5-Oxoazelaic acid
Formula: C₉H₁₄O₅
Número MDL: MFCD00075542
Núm. CAS: 57822-06-7

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (GC) min. 96.0 %
Purity (Neutralization titration) min. 96.0 %
Melting point 107.0 to 113.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR