(S)-(+)-5-Oxotetrahydrofuran-2-carboxylic acid ≥98.0% (by GC, titration analysis)
Proveedor: TCI
|
Atención
|
O0276-1G
O0276-5G
TCIAO0276-1GEA
38
EUR
TCIAO0276-1G
TCIAO0276-5G
(S)-(+)-5-Oxotetrahydrofuran-2-carboxylic acid ≥98.0% (by GC, titration analysis)
(S)-(+)-5-Oxotetrahydrofuran-2-carboxylic acid
Formula:
C₅H₆O₄ |
Número MDL:
MFCD00064516 Núm. CAS: 21461-84-7 |
Resultados test especificaciones
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Optical purity (GC) | min. 98.0 ee% |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 71.0 to 75.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...