Potassium hydrogen monopersulphate >∼45% KHSO5
Proveedor: TCI
|
Peligro
|
Sinónimos:
KMPS, Potassium peroxomonosulphate, Oxone, PMPS, Potassium monopersulphate, Potassium monopersulphate triple salt
O0310-25G
O0310-500G
TCIAO0310-25GEA
16
EUR
TCIAO0310-25G
TCIAO0310-500G
Potassium hydrogen monopersulphate >∼45% KHSO5
Potassium hydrogen monopersulphate
Formula:
2KHSO₅·KHSO₄·K₂SO₄ MW: 614,76 g/mol |
Número MDL:
MFCD00040551 Núm. CAS: 37222-66-5 UN: 3260 ADR: 8,II |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (Iodometric Titration) | 42.0 to 48.0 % (as KHSO5) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...