[(Oxido)phenyl(trifluoromethyl)-λ-4-sulphanylidene]dimethylammonium tetrafluoroborate 98,0 HPLC, N

Proveedor: TCI
Peligro

Sinónimos: Shibata Reagent I

O0367-1G O0367-200MG
TCIAO0367-1GEA 413 EUR
TCIAO0367-1G TCIAO0367-200MG
[(Oxido)phenyl(trifluoromethyl)-λ-4-sulphanylidene]dimethylammonium tetrafluoroborate 98,0 HPLC, N
[(Oxido)phenyl(trifluoromethyl)-lambda-4-sulphanylidene]dimethylammonium tetrafluoroborate
Formula: C₉H₁₁BF₇NOS
MW: 325.05 g/mol
Núm. CAS: 1046786-08-6
UN: 1759
ADR: 8 III

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR