O-[2-Oxo-1(2H)-pyridyl]-N,N,N',N'-tetramethyluronium tetrafluoroborate ≥98.0% (by HPLC, total nitrogen)
Proveedor: TCI
|
Peligro
|
Sinónimos:
O-(2-Oxo-1(2H)pyridyl)-N,N,N',N'-tetramethyluronium tetrafluoroborate, O-(1,2-Dihydro-2-oxo-1-pyridyl)-N,N,N',N'-tetramethyluronium Tetrafluoroborate, TPTU
O0390-5G
O0390-1G
TCIAO0390-5GEA
56
EUR
TCIAO0390-5G
TCIAO0390-1G
O-[2-Oxo-1(2H)-pyridyl]-N,N,N',N'-tetramethyluronium tetrafluoroborate ≥98.0% (by HPLC, total nitrogen)
O-[2-Oxo-1(2H)-pyridyl]-N,N,N',N'-tetramethyluronium tetrafluoroborate
Formula:
C₁₀H₁₆BF₄N₃O₂ MW: 297,06 g/mol Temperatura de almacenaje: Nevera |
Número MDL:
MFCD00075475 Núm. CAS: 125700-71-2 UN: 1759 ADR: 8,III |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Melting point | 135.0 to 139.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...