3-Oxocyclobutanecarboxylic acid ≥97.0% (by GC, titration analysis)

Proveedor: TCI
Atención

O0392-5G O0392-1G
TCIAO0392-5GEA 104 EUR
TCIAO0392-5G TCIAO0392-1G
3-Oxocyclobutanecarboxylic acid ≥97.0% (by GC, titration analysis)
3-Oxocyclobutanecarboxylic acid
Formula: C5H6O3
MW: 114,10 g/mol
Número MDL: MFCD00100900
Núm. CAS: 23761-23-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=927a793e1f5f301c20bd227387d0e6d4f0297f4a&vid=927a793e1f5f301c20bd227387d0e6d4f0297f4a&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (GC) min. 97.0 %
Purity (Neutralization titration) min. 97.0 %
Melting point 68.0 to 72.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR