Olanzapine ≥98.0% (by GC, titration analysis)

Proveedor: TCI
Peligro

O0393-1G O0393-5G
TCIAO0393-1GEA 55 EUR
TCIAO0393-1G TCIAO0393-5G
Olanzapine ≥98.0% (by GC, titration analysis)
Olanzapine
Formula: C₁₇H₂₀N₄S
Temperatura de almacenaje: Nevera
Número MDL: MFCD00866702
Núm. CAS: 132539-06-1
UN: 2811
ADR: 6.1,III
Índice Merck: 14,06822
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=eb1f7c3409234bac85aa3a431dda19a2461301b8&vid=eb1f7c3409234bac85aa3a431dda19a2461301b8&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance Light yellow to Amber to Dark green powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 192.0 to 196.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR