Ornidazole ≥98.0% (by HPLC, titration analysis)
Proveedor: TCI
|
Atención
|
Sinónimos:
1-(3-Chloro-2-hydroxypropyl)-2-methyl-5-nitroimidazole, 1-Chloro-3-(2-methyl-5-nitro-1H-imidazol-1-yl)propan-2-ol
O0417-5G
O0417-25G
TCIAO0417-5GEA
41
EUR
TCIAO0417-5G
TCIAO0417-25G
Ornidazole ≥98.0% (by HPLC, titration analysis)
Ornidazole
Formula:
C₇H₁₀ClN₃O₃ MW: 219,63 g/mol |
Número MDL:
MFCD00057960 Núm. CAS: 16773-42-5 EINECS: 240-826-0 Índice Merck: 14,06872 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 90.0 to 94.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...