Omeprazole sulphide ≥98.0% (by HPLC, titration analysis)
Proveedor: TCI
Sinónimos:
5-Methoxy-2-[[(4-methoxy-3,5-dimethyl-2-pyridyl)methyl]thio]benzimidazole
O0437-200MG
O0437-1G
TCIAO0437-200MGEA
53
EUR
TCIAO0437-200MG
TCIAO0437-1G
Omeprazole sulphide ≥98.0% (by HPLC, titration analysis)
Omeprazole sulphide
Formula:
C₁₇H₁₉N₃O₂S MW: 329,42 g/mol |
Número MDL:
MFCD00869021 Núm. CAS: 73590-85-9 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 122.0 to 126.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...