2,2'-Oxydiacetyl chloride ≥97.0% (by GC, titration analysis)
Proveedor: TCI
|
Peligro
|
O0459-5G
O0459-25G
TCIAO0459-5GEA
34
EUR
TCIAO0459-5G
TCIAO0459-25G
2,2'-Oxydiacetyl chloride ≥97.0% (by GC, titration analysis)
2,2'-Oxydiacetyl chloride
Formula:
C₄H₄Cl₂O₃ Pto. Ebullición: 57°C |
Número MDL:
MFCD00134471 Núm. CAS: 21062-20-4 EINECS: 244-186-3 UN: 3265 ADR: 8,II |
|
Resultados test especificaciones
Appearance | Colorless to Light orange to Yellow clear liquid |
Purity (GC) | min. 97.0 % |
Purity (Argentmetric Titration) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...