Ocaperidone ≥98.0% (by HPLC)
Proveedor: TCI
Sinónimos:
3-[2-[4-(6-Fluorobenzo[d]isoxazol-3-yl)piperidin-1-yl]ethyl]-2,9-dimethyl-4H-pyrido[1,2-a]pyrimidin-4-one
O0498-100MG
O0498-25MG
TCIAO0498-100MGEA
351
EUR
TCIAO0498-100MG
TCIAO0498-25MG
Ocaperidone ≥98.0% (by HPLC)
Ocaperidone
Formula:
C₂₄H₂₅FN₄O₂ MW: 420,49 g/mol |
Número MDL:
MFCD00866668 Núm. CAS: 129029-23-8 |
Resultados test especificaciones
Appearance | White to Light yellow powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Elemental analysis (Nitrogen) | 12.80 to 13.80 % |
Melting point | 180.0 to 184.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...