Sodium 4-phenylbutyrate ≥98.0% (by HPLC, titration analysis)

Proveedor: TCI

O0511-1G O0511-5G
TCIAO0511-1GEA 45 EUR
TCIAO0511-1G TCIAO0511-5G
Sodium 4-phenylbutyrate ≥98.0% (by HPLC, titration analysis)
Sodium 4-phenylbutyrate
Formula: C₁₀H₁₁O₂·Na
Número MDL: MFCD00800247
Núm. CAS: 1716-12-7

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Ion exchange titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR