3-Oxo-4-aza-5alpha-androst-1-ene-17beta-carboxylic acid ≥98.0% (by HPLC, titration analysis)
Proveedor: TCI
Sinónimos:
(4aR,4bS,6aS,7S,9aS,9bS,11aR)-4a,6a-Dimethyl-2-oxo-2,4a,4b,5,6,6a,7,8,9,9a,9b,10,11,11a-tetradecahydro-1H-indeno[5,4-f]quinoline-7-carboxylic acid
O0515-1G
O0515-5G
TCIAO0515-1GEA
48
EUR
TCIAO0515-1G
TCIAO0515-5G
3-Oxo-4-aza-5alpha-androst-1-ene-17beta-carboxylic acid ≥98.0% (by HPLC, titration analysis)
3-Oxo-4-aza-5alpha-androst-1-ene-17beta-carboxylic acid
Formula:
C₁₉H₂₇NO₃ MW: 317.43 g/mol Temperatura de almacenaje: Congelador |
Número MDL:
MFCD08063794 Núm. CAS: 104239-97-6 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Specific rotation [a]20/D | -3.0 to -6.0 deg (C=1, pyridine) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...