Sodium 2,4,8,10-Tetra-tert-butyl-12H-dibenzo[d,g][1,3,2]dioxaphosphocin-6-olate 6-Oxide ≥98.0% (by HPLC, titration analysis)

Proveedor: TCI
Atención

O0527-25G O0527-5G
TCIAO0527-25GEA 130 EUR
TCIAO0527-25G TCIAO0527-5G
Sodium 2,4,8,10-Tetra-tert-butyl-12H-dibenzo[d,g][1,3,2]dioxaphosphocin-6-olate 6-Oxide ≥98.0% (by HPLC, titration analysis)
Sodium 2,4,8,10-Tetra-tert-butyl-12H-dibenzo[d,g][1,3,2]dioxaphosphocin-6-olate 6-Oxide
Formula: C₂₉H₄₂NaO₄P
Número MDL: MFCD01748150
Núm. CAS: 85209-91-2
EINECS: 286-344-4
UN: 3077
ADR: 9,III

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR