2-Oxohexahydro-2H-3,5-methanocyclopenta[b]furan-6-yl methacrylate ≥98.0% (por GC)
Proveedor: TCI
|
Atención
|
Sinónimos:
Methacrylic acid 2-Oxohexahydro-2H-3,5-methanocyclopenta[b]furan-6-yl ester, 5-Methacroyloxy-2,6-norbornane Carbolactone
O0536-5G
O0536-25G
TCIAO0536-5GEA
68
EUR
TCIAO0536-5G
TCIAO0536-25G
2-Oxohexahydro-2H-3,5-methanocyclopenta[b]furan-6-yl methacrylate ≥98.0% (por GC)
2-Oxohexahydro-2H-3,5-methanocyclopenta[b]furan-6-yl methacrylate
Formula:
C₁₂H₁₄O₄ MW: 222,24 g/mol |
Número MDL:
MFCD31715034 Núm. CAS: 254900-07-7 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 100.0 to 104.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...