4-Phenylazobenzoyl chloride ≥98.0% (by GC, titration analysis)
Proveedor: TCI
|
Peligro
|
Sinónimos:
Azobenzene-4-carbonyl Chloride, Azoyl Chloride
P0144-1G
P0144-5G
TCIAP0144-1GEA
68
EUR
TCIAP0144-1G
TCIAP0144-5G
4-Phenylazobenzoyl chloride ≥98.0% (by GC, titration analysis)
4-Phenylazobenzoyl chloride
Formula:
C₁₃H₉ClN₂O MW: 244,68 g/mol |
Número MDL:
MFCD00041722 Núm. CAS: 104-24-5 EINECS: 203-188-4 UN: 3261 ADR: 8,III |
Resultados test especificaciones
Appearance | Light yellow to Brown powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Argentmetric Titration) | min. 98.0 % |
Melting point | 93.0 to 97.0 deg-C |
Solubility in hot 2-Propanol | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...