Phenylpyruvic acid ≥93.0% (by GC, titration analysis)
Proveedor: TCI
|
Atención
|
Sinónimos:
alpha-Keto-DL-phenylalanine
P0154-25G
P0154-5G
TCIAP0154-25GEA
177
EUR
TCIAP0154-25G
TCIAP0154-5G
Phenylpyruvic acid ≥93.0% (by GC, titration analysis)
Phenylpyruvic acid
Formula:
C₉H₈O₃ MW: 164,16 g/mol Temperatura de almacenaje: Congelador |
Número MDL:
MFCD00002589 Núm. CAS: 156-06-9 EINECS: 205-847-1 |
Resultados test especificaciones
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 93.0 % |
Purity (Neutralization titration) | min. 93.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...