2-Phenylphenol sodium salt tetrahydrate ≥80.0% (by titrimetric analysis)

Proveedor: TCI
Peligro

P0202-25G P0202-500G
TCIAP0202-25GEA 16 EUR
TCIAP0202-25G TCIAP0202-500G
2-Phenylphenol sodium salt tetrahydrate ≥80.0% (by titrimetric analysis)
2-Phenylphenol sodium salt tetrahydrate
Formula: C₁₂H₁₀O
Pto. de fusión: 78°C
Número MDL: MFCD00002209
Núm. CAS: 6152-33-6
EINECS: 205-055-6
UN: 1759
ADR: 8,II
Índice Merck: 14,07304

Order Now

Resultados test especificaciones

Appearance White to Light yellow to Light orange powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 80.0 % (as Anhydride)
Water 25.0 to 30.0 %

ESPECIFICACIONES

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR