2-Ethylhexyl phosphate (mono and di ester mixture)
Proveedor: TCI
|
Peligro
|
P0261-25G
P0261-500G
TCIAP0261-25GEA
16
EUR
TCIAP0261-25G
TCIAP0261-500G
2-Ethylhexyl phosphate (mono and di ester mixture)
2-Ethylhexyl phosphate (mono and di ester mixture)
Número MDL:
MFCD00135884 Núm. CAS: 12645-31-7 EINECS: 235-741-0 UN: 3265 ADR: 8,III |
Resultados test especificaciones
Appearance | Colorless to Light yellow to Light orange clear liquid |
Assay of mono ester | 30.0 to 45.0 % |
Assay of Diester | 50.0 to 70.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...