Phthalyl-DL-glutamic acid ≥98.0% (by HPLC, titration analysis)

Proveedor: TCI

Sinónimos: Phthaloyl-DL-glutamic Acid

P0338-25G P0338-5G
TCIAP0338-25GEA 218 EUR
TCIAP0338-25G TCIAP0338-5G
Phthalyl-DL-glutamic acid ≥98.0% (by HPLC, titration analysis)
Phthalyl-DL-glutamic acid
Formula: C₁₃H₁₁NO₆
MW: 277,23 g/mol
Número MDL: MFCD00023059
Núm. CAS: 6349-98-0
EINECS: 228-754-8

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 189.0 to 193.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR