Sodium-1-propanesulphonate ≥98.0% (by titrimetric analysis)
Proveedor: TCI
Sinónimos:
Propanesulphonic acid sodium salt, 1-Propanesulphonic acid Sodium Salt, Propanesulphonic acid Sodium Salt, IPC-ALKS-3
P0347-25G
P0347-5G
TCIAP0347-25GEA
423
EUR
TCIAP0347-25G
TCIAP0347-5G
Sodium-1-propanesulphonate ≥98.0% (by titrimetric analysis)
Sodium-1-propanesulphonate
Formula:
C₃H₈O₃S MW: 146.14 g/mol |
Número MDL:
MFCD00062546 Núm. CAS: 14533-63-2 EINECS: 238-564-7 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (Ion exchange titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...