2,3-Dihidroftalazina-1,4-diona ≥98.0% (by HPLC, titration analysis)
Proveedor: TCI
Sinónimos:
2,3-Dihydrophthalazine-1,4-dione
P0354-25G
P0354-500G
TCIAP0354-25GEA
19
EUR
TCIAP0354-25G
TCIAP0354-500G
2,3-Dihidroftalazina-1,4-diona ≥98.0% (by HPLC, titration analysis)
2,3-Dihidroftalazina-1,4-diona
Formula:
C₈H₆N₂O₂ MW: 162,15 g/mol |
Número MDL:
MFCD00006888 Núm. CAS: 1445-69-8 EINECS: 215-893-4 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Solubility in hot Acetic acid | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...