Phenylthiohydantoin-methionine ≥98.0% (by HPLC, total nitrogen)

Proveedor: TCI

Sinónimos: Phenylthiohydantoin-methionine, 5-[2-(Methylthio)ethyl]-3-phenyl-2-thiohydantoin, PTH-methionine, 5-(2-(Methylthio)ethyl)-3-phenyl-2-thioxoimidazolidin-4-one

P0366-1G P0366-100MG
TCIAP0366-1GEA 86 EUR
TCIAP0366-1G TCIAP0366-100MG
Phenylthiohydantoin-methionine ≥98.0% (by HPLC, total nitrogen)
Phenylthiohydantoin-methionine
Formula: C₁₂H₁₄N₂OS₂
MW: 266,38 g/mol
Número MDL: MFCD06858379
Núm. CAS: 4370-90-5
EINECS: 224-464-0

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 131.0 to 135.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR