Phenylthiohydantoin-tryptophan ≥98.0% (by HPLC, total nitrogen)
Proveedor: TCI
Sinónimos:
5-(3-Indolylmethyl)-3-phenyl-2-thiohydantoin, PTH-tryptophan
P0378-1G
P0378-100MG
TCIAP0378-1GEA
142
EUR
TCIAP0378-1G
TCIAP0378-100MG
Phenylthiohydantoin-tryptophan ≥98.0% (by HPLC, total nitrogen)
Phenylthiohydantoin-tryptophan
Formula:
C₁₈H₁₅N₃OS MW: 321,40 g/mol |
Número MDL:
MFCD06858416 Núm. CAS: 5789-24-2 EINECS: 227-323-1 |
Resultados test especificaciones
Appearance | Light yellow to Yellow to Orange powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Melting point | 181.0 to 185.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...