Polyethyleneimine ∼30% in water

Proveedor: TCI
Peligro

Sinónimos: Polyaziridine, Corcat, Polyethylenimine, Polymin P, Polyaziridine Mw 25000, Aziridine, PEI, Polyethylenimine (branched), Ethyleneimine Polymer, Poly(ethyleneimine)

P0381-500G P0381-25G
TCIAP0381-500GEA 43 EUR
TCIAP0381-500G TCIAP0381-25G
Polyethyleneimine ∼30% in water
Polyethyleneimine
Formula: (CH2CH2NH)n
Número MDL: MFCD00084427
Núm. CAS: 9002-98-6

Order Now

Resultados test especificaciones

Appearance Colorless to Almost colorless clear liquid
Total Nitrogen 9.2 to 10.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR