Trioleyl phosphite (so called) ≥98.0% (by titrimetric analysis)

Proveedor: TCI

Sinónimos: Phosphorous Acid Trioleyl Ester

P0390-25ML P0390-500ML
TCIAP0390-25MLEA 16 EUR
TCIAP0390-25ML TCIAP0390-500ML
Trioleyl phosphite (so called) ≥98.0% (by titrimetric analysis)
Trioleyl phosphite
Formula: C₅₄H₁₀₅O₃P
MW: 833,40 g/mol
Pto. de inflamación: 225°C
Número MDL: MFCD00059452
Núm. CAS: 13023-13-7

Order Now

Resultados test especificaciones

Appearance Colorless to Light orange to Yellow clear liquid
Purity (Iodometric Titration) min. 98.0 % (as Trioleyl Phosphite)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR