Polivinilpirrolidona, M.W. ∼ 360000 K 90

Proveedor: TCI

Sinónimos: Povidone, Polyvinylpyrrolidon, Polyvidone, PVP

P0473-25G P0473-500G
TCIAP0473-25GEA 17 EUR
TCIAP0473-25G TCIAP0473-500G
Polivinilpirrolidona, M.W. ∼ 360000 K 90
Polyvinylpyrrolidone
Formula: (C₆H₉NO)n
Número MDL: MFCD01076626
Núm. CAS: 9003-39-8
Índice Merck: 14,07697

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Total Nitrogen 12.0 to 12.8 %
Water max. 5.0 %
K value 84.0 to 95.0

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR