Ácido 2,3,4-trihidroxibenzoico ≥98.0% (by titrimetric analysis)
Proveedor: TCI
|
Atención
|
P0571-1G
P0571-5G
TCIAP0571-1GEA
18
EUR
TCIAP0571-1G
TCIAP0571-5G
Ácido 2,3,4-trihidroxibenzoico ≥98.0% (by titrimetric analysis)
Ácido 2,3,4-trihidroxibenzoico
Formula:
C₇H₆O₅ MW: 170,12 g/mol |
Número MDL:
MFCD00002447 Núm. CAS: 610-02-6 EINECS: 210-205-9 |
Resultados test especificaciones
Appearance | White to Light gray to Light orange powder to crystal |
Purity (HPLC) | min. 95.0 area% |
Purity (Neutralization titration) | min. 98.0 % (calcd.on anh.substance) |
Water | max. 5.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...