Phenylglyoxal monohydrate ≥98.0% (por GC)
Proveedor: TCI
|
Atención
|
P0652-25G
P0652-5G
TCIAP0652-25GEA
859
EUR
TCIAP0652-25G
TCIAP0652-5G
Phenylglyoxal monohydrate ≥98.0% (por GC)
Phenylglyoxal monohydrate
Formula:
C₈H₆O₂·H₂O |
Número MDL:
MFCD00149499 Núm. CAS: 1075-06-5 EINECS: 214-036-1 |
Resultados test especificaciones
Appearance | White powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 76.0 to 91.0 deg-C |
Water | 11.0 to 13.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...