Polyethylene glycol monooleyl ether n=∼50

Proveedor: TCI

P0716-25G P0716-500G
TCIAP0716-25GEA 16 EUR
TCIAP0716-25G TCIAP0716-500G
Polyethylene glycol monooleyl ether n=∼50
Polyethylene glycol monooleyl ether
Formula: HO(CH₂CH₂O)nC₁₈H₃₅
Número MDL: MFCD00074369
Núm. CAS: 9004-98-2
EINECS: 500-016-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5ebd9deaba8d9f29b03cee9a0450b0908efead64&vid=8f5c8c8e73507cd6545c98f23dfbf630c9b4fe55&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white lump
The average value of n 40.0 to 65.0
Acid Value max. 1.0

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR