Pentaerythritol tetrakis [3-(3,5-di-tert-butyl-4-hydroxyphenyl)propionate] ≥95.0% (by HPLC)
Proveedor: TCI
Sinónimos:
Tetrakis[methylene-3-(3,5-di-tert-butyl-4-hydroxyphenyl)propionate]methane, Antioxidant 1010, 2,2-Bis(((3-(3,5-di-tert-butyl-4-hydroxyphenyl)propanoyl)oxy)methyl)propane-1,3-diyl bis(3-(3,5-di-tert-butyl-4-hydroxyphenyl)propanoate)
P0932-25G
P0932-500G
P0932-100G
TCIAP0932-25GEA
17
EUR
TCIAP0932-25G
TCIAP0932-500G
TCIAP0932-100G
Pentaerythritol tetrakis [3-(3,5-di-tert-butyl-4-hydroxyphenyl)propionate] ≥95.0% (by HPLC)
Pentaerythritol tetrakis [3-(3,5-di-tert-butyl-4-hydroxyphenyl)propionate]
Formula:
C₇₃H₁₀₈O₁₂ |
Número MDL:
MFCD00059345 Núm. CAS: 6683-19-8 EINECS: 229-722-6 |
|
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 95.0 area% |
Melting point | 114.0 to 121.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...